Skip to Content
Merck
All Photos(1)

Key Documents

03701

Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

≥97.0% (KT)

Synonym(s):

Tetrasodium EDTA, EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€48.80
1 KG
€110.00

€48.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€48.80
1 KG
€110.00

About This Item

Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COONa)2 · xH2O
CAS Number:
Molecular Weight:
380.17 (anhydrous basis)
Beilstein:
3861753
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
powder

€48.80


Estimated to ship on28 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

≥97.0% (KT)

form

powder

functional group

amine

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.4Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;1H2/q;4*+1;/p-4

InChI key

GXFFSKMBMYHTAE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

EDTA is an inhibitor of metalloproteases, at effective concentrations of 1-10 mM. EDTA acts as a chelator of the zinc ion in the active site of metalloproteases, and can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases. For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used. The optimal concentration is 1.5 mg per ml of blood.
Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.

Preparation Note

Soluble in water at room temperature up to 1.45 M. The pH of this solution will be approximately 10 to 11 and will be rather viscous.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service