![Ethylenediaminetetraacetic acid tetrasodium salt hydrate purum, ≥80% C10H12N2Na4O8 basis](/deepweb/assets/sigmaaldrich/product/structures/414/256/966cad30-6281-4983-b42c-bd9d86bad78e/640/966cad30-6281-4983-b42c-bd9d86bad78e.png)
Ethylenediaminetetraacetic acid tetrasodium salt hydrate
![Ethylenediaminetetraacetic acid tetrasodium salt hydrate purum, ≥80% C10H12N2Na4O8 basis](/deepweb/assets/sigmaaldrich/product/structures/414/256/966cad30-6281-4983-b42c-bd9d86bad78e/640/966cad30-6281-4983-b42c-bd9d86bad78e.png)
Synonym(s):
EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate hydrate
Linear Formula:
(NaO2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O
Products
Brand
Biological Source
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reagent Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing