Skip to Content
Merck
All Photos(1)

Key Documents

03553

Supelco

Griess′ reagent for nitrite

for microscopy

Synonym(s):

Griess-Ilosvays Reagent for nitrate, Lunge’s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

grade

for microscopy

form

liquid

impurities

2-naphthylamine
acetic acid

refractive index

n20/D 1.353

density

1.037 g/mL at 20 °C

suitability

suitable for microscopy

application(s)

food and beverages
general analytical

General description

The Griess reagent is used for the quantitative analysis of nitrite ions. Present nitrite (as well as nitrate) ions will form a pink diazo dye by diazonium coupling reaction with N-(1-Naphthyl)ethylenediamine. The concentration of nitrite (or nitrate) ions is proportional to the color intensity of the resulting red solution.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su Young Chae et al.
Biomaterials, 25(5), 843-850 (2003-11-12)
Pancreatic islets and insulinoma cells are particularly vulnerable to serious damage by cytotoxic nitric oxide (NO) and/or oxidative stress, most probably due to their low expression levels of antioxidant enzymes. This cellular damage has been regarded as one of major
Anne M Sanders et al.
BMC cancer, 10, 121-121 (2010-04-02)
Spontaneous Regression/Complete Resistant (SR/CR) mice are a colony of cancer-resistant mice that can detect and rapidly destroy malignant cells with innate cellular immunity, predominately mediated by granulocytes. Our previous studies suggest that several effector mechanisms, such as perforin, granzymes, or
Qing-Li Wu et al.
Journal of immunology (Baltimore, Md. : 1950), 182(11), 6771-6778 (2009-05-21)
The Emu-TCL1 transgenic mouse spontaneously develops a CD5(+) B cell lymphoproliferative disorder similar to human chronic lymphocytic leukemia (CLL). Given the ineffectual T cell antitumor responses in this mouse model of CLL, we sought to determine whether combined treatment with
A Campos-Neto et al.
Journal of immunology (Baltimore, Md. : 1950), 160(5), 2037-2041 (1998-03-14)
It has been proposed that the induction of cellular immunity and resistance to intracellular pathogens is dependent upon CD40 ligand (CD40L). In the present study we show that this proposal is not ubiquitously supported. Mice genetically deficient in CD40L (CD40LKO)
Lei Qiu et al.
Immunology, 128(1 Suppl), e325-e334 (2008-11-20)
Using two-dimensional sodium dodecyl sulphate-polyacrylamide gel electrophoresis, we found that copper/zinc superoxide dismutase (Cu/Zn-SOD, SOD-1) was induced in constructed CCR5 stably transfected HEK 293 cells, but not in mock cells, treated with CCL5. CCL5-induced SOD-1 expression was also confirmed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service