Skip to Content
Merck
All Photos(1)

Key Documents

01260595

Coumarin

primary reference standard

Synonym(s):

1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€242.00

€242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€242.00

About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein:
383644
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€242.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

vapor pressure

0.01 mmHg ( 47 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

bp

298 °C (lit.)

mp

68-73 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

SMILES string

O=C1Oc2ccccc2C=C1

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: cinnamomum curcuma hedera humulus melissa mentha salvia sambucus zingiber

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack Cazes
Encyclopedia of Chromatography, 206-206 (2001)
The pharmacology, metabolism, analysis, and applications of coumarin and coumarin-related compounds.
D Egan et al.
Drug metabolism reviews, 22(5), 503-529 (1990-01-01)
Shigehiro Sumiya et al.
The journal of physical chemistry. A, 117(7), 1474-1482 (2013-01-25)
A coumarin-amide-dipicolylamine linkage (L) was synthesized and used as a fluorescent receptor for metal cations in water. The receptor dissolved in water with neutral pH shows almost no fluorescence due to the photoinduced electron transfer (PET) from the amide and
B G Lake
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(4), 423-453 (1999-07-27)
The metabolism, toxicity and results of tests for carcinogenicity have been reviewed with respect to the safety for humans of coumarin present in foodstuffs and from fragrance use in cosmetic products. Coumarin is a natural product which exhibits marked species
Shota Morimoto et al.
Chemical communications (Cambridge, England), 49(18), 1811-1813 (2013-01-26)
A photo-switchable fluorescent flagging approach has been developed to identify photoaffinity-labeled peptides in target protein. Upon photochemical release of the ligand, the protein was newly modified with a coumarin in place of the previously attached biotin. It allowed us to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service