Skip to Content
Merck
All Photos(1)

Key Documents

416541

Sigma-Aldrich

Coumarin 7

98%

Synonym(s):

3-(2-Benzimidazolyl)-7-(diethylamino)coumarin

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€72.80

€72.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€72.80

About This Item

Empirical Formula (Hill Notation):
C20H19N3O2
CAS Number:
Molecular Weight:
333.38
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€72.80


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

234-237 °C (lit.)

λmax

438 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)c3nc4ccccc4[nH]3

InChI

1S/C20H19N3O2/c1-3-23(4-2)14-10-9-13-11-15(20(24)25-18(13)12-14)19-21-16-7-5-6-8-17(16)22-19/h5-12H,3-4H2,1-2H3,(H,21,22)

InChI key

GOLORTLGFDVFDW-UHFFFAOYSA-N

General description

Coumarin is also known as benzoαpyrone. Coumarin 7 is a laser dye that is usually found in the nature. Its derivatives are used in medicine and chemistry. It is present in plants in the form of glucosides.[1] Coumarin acts as an antimutagen as it controls the mutagenic effects of several chemicals like aflatoxin B1and physical agents like ultraviolet light.[2]

Application

Suitable as a laser dye

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisi Xie et al.
ACS nano, 14(3), 2880-2893 (2020-03-04)
Micro/nanorobots have been extensively explored as a tetherless small-scale robotic biodevice to perform minimally invasive interventions in hard-to-reach regions. Despite the emergence of versatile micro/nanorobots in recent years, matched in vivo development remains challenging, limited by unsatisfactory integration of core

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service