Recommended Products
vapor density
1.38 (21 °C, vs air)
Quality Level
Assay
≥99.998%
bp
-185.7 °C (lit.)
mp
-189.2 °C (lit.)
SMILES string
[Ar]
InChI
1S/Ar
InChI key
XKRFYHLGVUSROY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
filling pressure at 15°C; 11 bar, content (15°C, 1 bar) 11 L
Packaging
pressure tin
Other Notes
Sales restrictions may apply
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Clinical applications of the argon plasma coagulator.
Gastrointestinal endoscopy, 59(1), 81-88 (2004-01-15)
Clinical application of argon plasma coagulation in gastrointestinal endoscopy: has the time come to replace the laser?
Endoscopy, 33(4), 353-357 (2001-04-24)
American journal of respiratory cell and molecular biology, 51(3), 455-465 (2014-04-18)
Transplantation of mesenchymal stem cells (MSCs) holds great promise in the repair of cigarette smoke (CS)-induced lung damage in chronic obstructive pulmonary disease (COPD). Because CS leads to mitochondrial dysfunction, we aimed to investigate the potential benefit of mitochondrial transfer
Clinical & experimental ophthalmology, 43(2), 180-187 (2014-07-01)
Diabetic macular oedema (DMO) is the commonest cause of vision loss in people with diabetes. Laser photocoagulation can be effective in the treatment of DMO; however, its mechanism of action is still poorly understood. A better understanding of these mechanisms
Chemistry (Weinheim an der Bergstrasse, Germany), 20(33), 10397-10403 (2014-07-22)
Molecular assemblies that change photoluminescence color in response to thermal or mechanical stimulation without dissociation into the monomeric states in water are described herein. A dumbbell-shaped amphiphilic compound forms micellar molecular assemblies in water and exhibits yellow photoluminescence derived from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service