Skip to Content
Merck
All Photos(2)

Documents

769061

Sigma-Aldrich

Nitrogen

Messer® CANGas, 99.999%

Synonym(s):

Nitrogen gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
N2
CAS Number:
Molecular Weight:
28.01
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.97 (vs air)

Quality Level

description

Filling mass: 14g
Filling volume: 12L

Assay

99.999%

form

gas

bp

−196 °C (lit.)

mp

−210 °C (lit.)

SMILES string

N#N

InChI

1S/N2/c1-2

InChI key

IJGRMHOSHXDMSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nitrogen is a colourless, odourless, and asphyxiating gas that is slightly lighter than air. Nitrogen is used as a carrier gas in gas chromatography, an inert gas in chemical and pharmaceutical industries, and a zero gas for analytical applications, such as in the automotive industries. Nitrogen is also used as a component of gas mixtures for CO2 lasers. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service