Skip to Content
Merck
All Photos(1)

Key Documents

30-0399

Sigma-Aldrich

Terpineol

SAJ first grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥97.0%
bp:
213-218 °C (lit.)
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥97.0%

form

liquid

availability

available only in Japan

bp

213-218 °C (lit.)

density

0.934 g/mL at 20 °C (lit.)

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Chang Wang et al.
Sensors (Basel, Switzerland), 15(6), 14286-14297 (2015-06-20)
Highly sensitive H2 gas sensors were prepared using pure and Pt-loaded SnO2 nanoparticles. Thick film sensors (~35 μm) were fabricated that showed a highly porous interconnected structure made of high density small grained nanoparticles. Using Pt as catalyst improved sensor
Khaled Sebei et al.
Biological research, 48, 7-7 (2015-02-06)
In this paper, we have studied the essential oils chemical composition of the leaves of seven Eucalyptus species developed in Tunisia. Eucalyptus leaves were picked from trees growing in different arboretums in Tunisia. Choucha and Mrifeg arboretums located in Sedjnene
Yi-Fei Sun et al.
Nanoscale, 7(25), 11173-11181 (2015-06-11)
To date, most investigations of Ni-Fe bimetallic catalysts for solid oxide fuel cells (SOFCs) have focused on materials with micro-scale particle sizes, which severely restrict their catalytic activity. In this study, we fabricated a Ni- and/or Fe-doped A-site-deficient LaSrCrO3 perovskite
Dae-Yong Son et al.
Physical chemistry chemical physics : PCCP, 17(23), 15193-15200 (2015-05-21)
To understand the role of the dye/oxide interface, a model system using a nanocrystalline SnO2 and 3-hexyl thiophene based MK-2 dye is proposed. A thin interfacial TiO2 blocking layer (IBL) is introduced in between SnO2 and MK-2 and its effects
Han Fu et al.
Nanoscale research letters, 9(1), 631-631 (2014-12-19)
Fabricating functional compounds on substrates with complicated morphology has been an important topic in material science and technology, which remains a challenging issue to simultaneously achieve a high growth rate for a complex nanostructure with simple controlling factors. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service