Skip to Content
Merck
All Photos(7)

Key Documents

SAB5701095

Sigma-Aldrich

Anti- SIRT1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€241.00

€241.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB07131


Select a Size

Change View
100 μL
€241.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€241.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB07131

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human, mouse, rat

concentration

2.56 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT1(23411)

General description

This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Alternative splicing results in multiple transcript variants.

Immunogen

A synthetic peptide of human SIRT1

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service