Skip to Content
Merck
All Photos(1)

Key Documents

28-4640

Sigma-Aldrich

Sodium tungstate dihydrate

JIS special grade, 99.0-100.5

Synonym(s):

Tungstic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99.0-100.5
grade:
JIS special grade
form:
crystals
Pricing and availability is not currently available.

grade

JIS special grade

Assay

99.0-100.5

form

crystals

reaction suitability

reagent type: catalyst
core: tungsten

availability

available only in Japan

pH

9.0-11.5 (20 °C, 100 g/L)

mp

698 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delia Zafra et al.
FEBS letters, 587(3), 291-296 (2012-12-25)
Tungstate treatment ameliorates experimental diabetes by increasing liver glycogen deposition through an as yet unidentified mechanism. The signalling mechanism of tungstate was studied in CHOIR cells and primary cultured hepatocytes. This compound exerted its pro-glycogenic effects through a new G-protein-dependent
Gustavo S Guandalini et al.
Chemical research in toxicology, 24(4), 488-493 (2011-03-08)
Heavy metal tungsten alloys have replaced lead and depleted uranium in many munitions applications, due to public perception of these elements as environmentally unsafe. Tungsten materials left in the environment may become bioaccessible as tungstate, which might lead to population
Andrew R Osterburg et al.
Journal of immunotoxicology, 7(3), 174-182 (2010-02-25)
The potential for adverse health effects of using tungsten and its alloys in military munitions are an important concern to both civilians and the US military. The toxicological implications of exposure to tungsten, its alloys, and the soluble tungstate (Na(2)WO(4))
M Milagros Gómez-Gómez et al.
Talanta, 84(4), 1011-1018 (2011-05-03)
It is known that oral administration of sodium tungstate preserves the pancreatic beta cell function in diabetic rats. Healthy and streptozotocin-induced diabetic rats were treated with sodium tungstate for one, three or six weeks, after which the species of W
M Amigó-Correig et al.
Diabetes, obesity & metabolism, 13(3), 235-242 (2011-01-06)
Sodium tungstate is an anti-obesity drug targeting peripheral tissues. In vivo, sodium tungstate reduces body weight gain and food intake through increasing energy expenditure and lipid oxidation, but it also modulates hypothalamic gene expression when orally administered, raising the possibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service