Skip to Content
Merck
All Photos(1)

Key Documents

03-3075

Sigma-Aldrich

Boron trifluoride-methanol solution

CP

Synonym(s):

Boron trifluoride-methanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · MeOH
CAS Number:
Molecular Weight:
99.85
Beilstein:
3611499
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

CP

form

liquid

availability

available only in Japan

concentration

~51% boron trifluoride

density

1.203 g/cm3

SMILES string

CO.FB(F)F

InChI

1S/CH4O.BF3/c1-2;2-1(3)4/h2H,1H3;

InChI key

JBXYCUKPDAAYAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

83.3 °F

Flash Point(C)

28.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Hue-Beauvais et al.
Journal of animal science, 93(4), 1641-1655 (2015-05-29)
Alterations to the metabolic endocrine environment during early life are crucial to mammary gland development. Among these environmental parameters, the initial nutritional event after birth is the consumption of milk, which represents the first maternal support provided to mammalian newborns.
Leila Nejadsadeghi et al.
Molecular biology reports, 42(2), 363-372 (2014-10-01)
Plant cells often increase cold tolerance by reprogramming their genes expression which results in adjusted metabolic alternations, a process enhanced under cold acclimation. In present study, we assessed the changes of membrane fatty acid compositions along with physio-biochemical indices like
Shulin Xiang et al.
Journal of pineal research, 59(1), 60-69 (2015-04-11)
Chemotherapeutic resistance, particularly to doxorubicin (Dox), represents a major impediment to successfully treating breast cancer and is linked to elevated tumor metabolism and tumor over-expression and/or activation of various families of receptor- and non-receptor-associated tyrosine kinases. Disruption of circadian time
Tao Wang et al.
Molecular medicine reports, 12(2), 3200-3206 (2015-05-13)
The aim of the present study was to investigate the effects of dietary trans fatty acids in mice. Following the administration of a 0.5/100 g diet of trans-9 octadecenoic acid (EA), trans-11 vaccenic acid (TVA) or cis-9, trans-11 conjugated linoleic
Nancy Hachicho et al.
PloS one, 10(8), e0134755-e0134755 (2015-08-21)
Morphology and physiology of fish embryos undergo dramatic changes during their development until the onset of feeding, supplied only by endogenous yolk reserves. For obtaining an insight how these restructuring processes are reflected by body mass related parameters, dry weights

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service