Key Documents
ASMUVLPA1
Milli-Q
ech2o® ASM Lamp
Protects stored water against biofilm buildup in Milli-Q® IQ/IX storage tanks
Select a Size
1 EACH
€415.00
€415.00
Estimated to ship on16 April 2025
Request more information
Select a Size
Change View
1 EACH
€415.00
About This Item
UNSPSC Code:
41104200
NACRES:
NB.33
€415.00
Estimated to ship on16 April 2025
Request more information
Quality Level
feature
mercury-free UV LED
packaging
pkg of 1 unit
manufacturer/tradename
ech2o®
parameter
265 nm (wavelength)
compatibility
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
General description
The ECH2O® ASM lamp protects the integrity of the stored water with regular exposure to bactericidal UV light. This ECH2O® ASM lamp is a mercury-free UV LED (265 nm) that prevents the bacterial growth and biofilm formation on the inner surface of Milli-Q® IQ/IX storage tanks.
Application
It prevents the formation of biofilm and proliferation of bacteria inside the storage tank.
Legal Information
ECH2O is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany