Skip to Content
Merck
All Photos(1)

Key Documents

01-1350

Sigma-Aldrich

Acrylonitrile

SAJ first grade, ≥98.0%

Synonym(s):

Vinyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCN
CAS Number:
Molecular Weight:
53.06
Beilstein:
605310
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

vapor density

1.83 (vs air)

vapor pressure

86 mmHg ( 20 °C)

Assay

≥98.0%

autoignition temp.

897 °F

expl. lim.

17 %

availability

available only in Japan

refractive index

n20/D 1.391 (lit.)

bp

77 °C (lit.)

mp

−83 °C (lit.)

SMILES string

C=CC#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acrylonitrile is used in the manufacture of acrylic fibers, resins (acrylonitrile butadiene-styrene, styrene-acrylonitrile and others) and nitrile rubbers (butadiene-acrylonitrile).

Biochem/physiol Actions

An industrial carcinogen that is a multisite carcinogen in rats and possibly carcinogenic to humans.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Whysner et al.
Regulatory toxicology and pharmacology : RTP, 27(3), 217-239 (1998-08-07)
Acrylonitrile (ACN) exposure is associated with tumors in rat brain, Zymbal gland, and mammary gland. Adducts affecting base pairing were formed in isolated DNA exposed in vitro to the ACN metabolite cyanoethylene oxide (CNEO). DNA from liver, which is not
A Léonard et al.
Mutation research, 436(3), 263-283 (1999-06-04)
Acrylonitrile (AN) is an important intermediary for the synthesis of a variety of organic products, such as artificial fibres, household articles and resins. Although acute effects are the primary concern for an exposure to AN, potential genotoxic, carcinogenic and teratogenic
Acrylonitrile and human cancer--an overview.
D Coggon et al.
Scandinavian journal of work, environment & health, 24 Suppl 2, 81-82 (1998-08-26)
Philip Cole et al.
Regulatory toxicology and pharmacology : RTP, 52(3), 342-351 (2008-10-18)
Several retrospective cohort epidemiology studies evaluated a number of health outcomes in workers exposed to acrylonitrile (AN). The epidemiology studies included in this review have been published since 1970 and were identified through Ovid and MEDLINE retrieval services using search
N A Esmen
Scandinavian journal of work, environment & health, 24 Suppl 2, 63-70 (1998-08-26)
The reconstruction of worker exposures has been the mainstay of modem industrial epidemiologic studies. In most cases, the researchers are faced with the difficult dilemma created by the scarcity of the exposure measurement data vis-à-vis the demands for refined quantification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service