Skip to Content
Merck
All Photos(1)

Key Documents

8.00834

Sigma-Aldrich

Acrylonitrile

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

Acrylonitrile, Acrylic acid nitrile, Vinyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCN
CAS Number:
Molecular Weight:
53.06
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-466-5
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

124 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

480 °C

potency

78 mg/kg LD50, oral (Rat)

expl. lim.

2.8-28 % (v/v)

pH

6.0-7.5 (20 °C, 50 g/L in H2O)

bp

77.3 °C/1013 hPa

mp

-83.55 °C

transition temp

flash point -4.4 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

N#CC=C

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Application

  • Designing amphipathic surfaces: The use of interfacial energy in photopolymerization processes for creating amphipathic surfaces, enhancing applications in coatings and biomedical devices using acrylonitrile polymers (Curley et al., 2024).
  • Dental model printing technology: Atwal and Bhatnagar assessed the flexural strength of dental models fabricated using various 3D printing technologies, including those utilizing acrylonitrile-based resins, highlighting differences in mechanical properties (Atwal and Bhatnagar, 2024).
  • Rubber composite enhancements: Kruželák et al. examined the electrical conductivity, EMI absorption, and mechanical properties of rubber composites, incorporating acrylonitrile to improve performance in industrial applications (Kruželák et al., 2024).
  • Biopolymer blend characterization: Akbarian-Saravi et al. characterized the mechanical and biodegradation properties of NBR/biopolymer blends, with acrylonitrile enhancing the blend′s performance in various industrial and environmental applications (Akbarian-Saravi et al., 2024).
  • Electrode/electrolyte interface regulation: Chen et al. discussed constructing polymer molecules to regulate the electrode/electrolyte interface in lithium-metal batteries, utilizing acrylonitrile derivatives to enhance battery performance (Chen et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.805 - 0.807
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service