STREPWELL-RO
StreptaWell™ (transparent 96-well)
pkg of 5 plates (11664778001 [12 x 8 well strips and frame]), pkg of 15 plates (11734776001 [96 wells])
Synonym(s):
streptavidin-coated microplates
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
( streptavidin-coated)
packaging
pkg of 15 plates (11734776001 [96 wells])
pkg of 5 plates (11664778001 [12 x 8 well strips and frame])
manufacturer/tradename
Roche
suitability
suitable for immunoassay
storage temp.
2-8°C
Related Categories
General description
Transparent, streptavidin-coated microplates:
- Blocked volume: >300μL
- SA-coated area: 300μL coating variance guaranteed
- Well geometry: C
Application
StreptaWell™ can be used for colorimetric and fluorescent protein immunoassays.
Streptavidin-coated 96-well plate (StreptaWell) has been used to bound biotinylated peptide substrate contained supernatant:during in vitro kinase assays and DNA synthesis assay.
Streptavidin-coated 96-well plate (StreptaWell) has been used to bound biotinylated peptide substrate contained supernatant:during in vitro kinase assays and DNA synthesis assay.
Quality
SA leaching (bleeding): <5ng/well
Other Notes
Total biotin binding capacity: 5ng/well, 20pmol/well (competition assays; 300μl wells)
Total binding capacity: 1.5μg/well (biotin-labeled antibodies)
Total binding capacity: 1.5μg/well (biotin-labeled antibodies)
For life science research only. Not for use in diagnostic procedures.
Legal Information
StreptaWell is a trademark of Roche
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of virology, 84(23), 12325-12335 (2010-09-24)
Genome replication is inefficient without processivity factors, which tether DNA polymerases to their templates. The vaccinia virus DNA polymerase E9 requires two viral proteins, A20 and D4, for processive DNA synthesis, yet the mechanism of how this tricomplex functions is
The Journal of biological chemistry, 277(35), 31768-31773 (2002-06-26)
Tie2 is an endothelial receptor tyrosine kinase that is required for both embryonic vascular development and tumor angiogenesis. There is considerable interest in understanding the mechanisms of Tie2 activation for therapeutic purposes. The recent solution of the Tie2 crystal structure
Small (Weinheim an der Bergstrasse, Germany), 7(22), 3193-3201 (2011-10-01)
Zeolite L nanocrystals, as inorganic host material containing hydrophobic fluorophore N,N'-bis(2,6-dimethylphenyl)perylene-3,4,9,10-tetracarboxylic diimide in the unidirectional channels, are developed as new labels for biosensor systems. The external surface of the particles is modified with carboxylic acid groups for conjugation to primary
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service