Skip to Content
Merck
All Photos(1)

Key Documents

ZKITRES01T0CSD

Milli-Q

Resistivity Boost & TOC for High-throughput Systems

For use with Milli-Q® HX 7000 SD water systems

Synonym(s):

Resistivity Boost & TOC Monitor for High-flow Systems

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€4,550.00

€4,550.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 KIT
€4,550.00

About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

€4,550.00


Check Cart for Availability

Request more information

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® HX

General description

Installed within the purification system, the resistivity booster ensures the correct resistivity water flows in the loop and into the reservoir.
The in-line TOC (total organic carbon) monitor verifies TOC levels of system water is <500 ppb.

Features and Benefits

  • The resistivity booster may be easily installed on the front of the water purification system.
  • The ergonomic and patented pack-locking mechanism allows for rapid replacement when needed.
  • TOC monitor saves time and money and avoids the need for duplicate assays or repeated instrument calibrations.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service