Key Documents
ZKITRES00SD
Milli-Q
Resistivity Kit for High-throughput Systems
For use with Milli-Q® HX 7000 SD water systems
Select a Size
1 KIT
€913.00
€913.00
Check Cart for Availability
Request more information
Select a Size
Change View
1 KIT
€913.00
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
€913.00
Check Cart for Availability
Request more information
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® HX
General description
Installed within the purification system, this resistivity monitor verifies the resistivity level of water in the loop.
Features and Benefits
  • The resistivity booster can be easily installed on the front of the water purification system.
  • The ergonomic and patented pack-locking mechanism allows for rapid replacement when needed.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany