YP12
Power cord
for mA400 Basic Power Supply, mA700 Essential Power Supply and mPAGE® Lux Curing Station
Synonym(s):
power cord
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105303
Recommended Products
Product Name
YP12 NA power cord, for mA400 Basic Power Supply, mA700 Essential Power Supply and mPAGE® Lux Curing Station
AC/DC input
125 V, 10 A
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
General description
This electrical power cord has a specific plug configuration to match electrical outlets used in North America. This cord has been validated to replace the cord supplied with the mA400 Power Supply, mA700 Power Supply and the mPAGE® Lux Curing Station.
Compatibility
Compatible with:
mA400 Basic Power Supply
mA700 Essential Power Supply
mPAGE® Lux Curing Station
mA400 Basic Power Supply
mA700 Essential Power Supply
mPAGE® Lux Curing Station
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service