Skip to Content
Merck
All Photos(1)

Key Documents

TC-1L-BK

Millipore

Bulk Reagent Diluents - ABO & RHD (Rh) Specificities

ABO Reagent Diluent

Synonym(s):

ABO Reagent Diluent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.47

Note: For further information on price, availability or to place an order, please contact us

antibody form

unpurified

Quality Level

form

liquid

packaging

pack of 1 L

manufacturer/tradename

BIOSCOT

concentration

8-12 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

shipped in

ambient

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 3

This Item
TC-10L-BKTC-50L-BK
storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

600

Quality Level

600

Quality Level

600

shipped in

ambient

shipped in

ambient

shipped in

ambient

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

form

liquid

form

liquid

form

liquid

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

Product formulation examined according to Annex IV of the EU IVD directive to support CE registration.

For In Vitro use only

Application

Bulk.
For dilution of ABO reagents.

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service