Skip to Content
Merck

Skip To

TL-PACK

Blood typing Bulk Reagent - ABO & RHD (Rh) Specificities

CORE Anti-A (clone BIRMA-1) Bulk Reagent plus Diluent Kit

Synonym(s):

Anti-A (clone BIRMA-1), Blood Grouping Reagent plus Diluent

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
BIRMA-1, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-
Note: For further information on price, availability or to place an order, please contact us

biological source

mouse

antibody form

unpurified

clone

BIRMA-1, monoclonal

form

liquid

packaging

kit of

concentration

30-50 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Colorant: Patent Blue Violet dye

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

Application

For use by Tube;Slide;Microplate techniques.
Reagent + Diluent Kit.

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
TL-PACK04053252024344

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service