Skip to Content
Merck
All Photos(1)

Key Documents

SX0395

Supelco

Sodium carbonate

anhydrous, powder, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Soda, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-838-8
grade:
ACS reagent
form:
powder
solubility:
212.5 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

meets USP/NF

Assay

≥99.5% dry basis

form

powder

impurities

≤0.01% Insoluble matter

loss

≤1.0% loss on heating, 285°C

color

white

pH

11.16 (25 °C, 4 g/L in H2O)

mp

851 °C (lit.)

solubility

212.5 g/L

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
silicate (as SiO2): ≤0.005%
sulfur compounds (as SO42-): ≤0.003%

cation traces

Ca: ≤0.03%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.005%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, powder, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Analysis Note

Assay (dried basis)99.5% minCalcium (Ca)0.03% maxChloride (Cl)0.001% maxColorWhiteFormPowderHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.01% maxIron (Fe)5ppm maxLoss on heating at 285°C1.0% maxMagnesium (Mg)0.005% maxPhosphate (PO4)0.001% maxPotassium (K)0.005% maxSilica (SiO2)0.005% maxSulfur compounds (as SO4)0.003% max

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service