Skip to Content
Merck
All Photos(1)

Key Documents

SLHNX13

Millipore

Millex Nylon syringe filter

pore size 0.45 μm, diam. 13 mm, non-sterile, hydrophilic

Synonym(s):

0.45 μm Nylon syringe filter, Millex-HN syringe filter, disposable syringe filter, syringe filter

Sign Into View Organizational & Contract Pricing

Select a Size

100 PKG
€309.00
1000 PKG
€2,060.00

€309.00


Check Cart for Availability


Select a Size

Change View
100 PKG
€309.00
1000 PKG
€2,060.00

About This Item

UNSPSC Code:
41104922
eCl@ss:
32031690
NACRES:
NB.24

€309.00


Check Cart for Availability

material

Nylon membrane
polypropylene housing

Quality Level

Agency

suitable for DIN 38407-42
suitable for EPA OTM-45

sterility

non-sterile

product line

Millex

feature

holdup volume≤ 15 μL (after air purge)
hydrophilic

manufacturer/tradename

Millex

parameter

10 bar max. inlet pressure (150 psi)
37 L/min flow rate
37 mL/min-cm2 water flow rate
45 °C max. temp.

technique(s)

HPLC: suitable
UV/Vis spectroscopy: suitable
analytical sample preparation: suitable

H

21 mm

diam.

13 mm

filtration area

0.8 cm2

volume

10 mL

color

purple

pore size

0.45 μm

input

sample type aqueous solution(s)
sample type organic solution(s)
sample type mild organic solution(s)

bubble point

≥2.2 bar, air with water at 23 °C

fitting

female Luer-Lok® inlet
male Luer outlet slip

application(s)

PFAS testing

shipped in

ambient

General description

Millex Syringe Filter, Nylon, Non-sterile are hydrophilic nylon membranes that are compatible with a wide range of solvents including aqueous and organic solutions.

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
Millex is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrophilic PTFE Millex® syringe filters ensure accurate dissolution testing and sample preparation for HPLC analysis.

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service