Skip to Content
Merck
All Photos(1)

Documents

SF-CMTI-2

Sigma-Aldrich

ESGRO® Complete Adapted C57/BL6 Mouse ES Cell Line

Mouse

Synonym(s):

BRUCE-4 ES Cells, BRUCE-4 ES cells, CMTI-2 Mouse ES Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.52

product name

ESGRO® Complete Adapted C57/BL6 Mouse ES Cell Line, The Embryonic Stem Cell Lines derived from murine strain C57/BL6J are selected for high targeting efficiency & proven germ line transmission.

biological source

mouse

manufacturer/tradename

Specialty Media

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)

shipped in

liquid nitrogen

General description

Embryonic Stem Cell Lines derived from murine strain C57/BL6J selected for high targeting efficiency and proven germ line transmission. Cells are supplied as 2.5 x 106 ES cells per vial (approximately 2.5 population doublings per passage, shipped frozen on dry ice) and are supplied at passage 12. This ES cell line has been pre-adapted to serum-free and feeder-free cell culture conditions and is intended for use with the ESGRO Complete cell culture system.

Cell Line Description

Embryonic Stem Cells

Application

Research Category
Stem Cell Research

Packaging

2.5x106 ea

Storage and Stability

Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture.

Legal Information

ESGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meifang Li et al.
Chinese medicine, 15, 83-83 (2020-08-11)
Huyang Yangkun Formula (HYYKF) was developed based on theory of traditional Chinese medicine as well as clinical experience and used to improve ovarian function of premature ovarian insufficiency (POI) patients. Transplantation of embryonic stem cells (ESCs) has great potential in
Jixue Li et al.
The CRISPR journal, 3(6), 470-486 (2020-11-05)
We describe a protocol for the precise integration of exogenous DNA into user-defined genomic loci in cultured cells. This strategy first introduces a promoter and a lox site to a specific location via a Cas9-induced double-strand break. Second, a gene
Baisakhi Mondal et al.
eLife, 9 (2020-04-17)
The mitotic deacetylase complex (MiDAC) is a recently identified histone deacetylase (HDAC) complex. While other HDAC complexes have been implicated in neurogenesis, the physiological role of MiDAC remains unknown. Here, we show that MiDAC constitutes an important regulator of neural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service