Skip to Content
Merck
All Photos(1)

Documents

SF001

Sigma-Aldrich

ESGRO Complete PLUS Clonal Grade Medium, 100ml

to enhance viability of mouse embryonic stem cells

Synonym(s):

Mouse ES Cell Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.81

Quality Level

100
300

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)

General description

ESGRO Complete PLUS Clonal Grade Medium is a defined serum-free medium provided with a selective GSK3β inhibitor to enhance viability of mouse embryonic stem (ES) cells and increased maintenance of pluripotency in the absence of serum and feeder cells. This medium is intended for mouse ES cell expansion and growth at clonal density in serum-free conditions. The mouse ES cells should maintain germ line competency.

Application

Suitable for ES cell derivation, transfection and selection, in addition to general cell passaging and growth.
Media can be used on feeder dependent/independent ES cell lines.

Important passaging information
Use of one of the below listed cell dissociation solutions is imperative while passaging ES cells using ESGRO Complete products. Standard trypsin protocols used to passage ES cells grown in serum containing media typically result in over trypsinization causing the cells to lift off the plates and form abnormal ball-like structures.
Recommended products include:
ESGRO Complete Accutase (Cat # SF006)
ESGRO Complete Trypsin Solution (proprietary) (Cat # SF007)
ESGRO Complete Enzyme-Free Dissociation Solution (proprietary) (Cat # SF009)

Components

1. ESGRO Complete Clonal Grade Medium: (Catalogue No. SF001-C) One (1) 100 mL bottle containing original clonal grade medium. Complete formulation containing self-renewal factors, no need to add LIF. pH 7.1-7.3

2. GSK3β Inhibitor Supplement: (Catalogue No. SF012-050) One (1) 50 µL vial containing selective GSK3β inhibitor.

Quality

QC PROTOCOL:

Low density plating assay:
1. Plate feeder independent early passage ES cells onto gelatin coated 10 cm plastic tissue culture dish at 1000 cells/10 cm dish in 10-20 mL of Clonal Grade Medium.
2. Change medium every 2-4 days.
3. Monitor for clonal growth with alkaline phosphatase stain.

Colony formation should occur over 4-6 days. The majority of colonies should be alkaline phosphatase positive and show no signs of differentiation.

Linkage

Replaces: SF001-100

Storage and Stability

• Upon receipt, store clonal grade medium and supplement in the dark at -20°C. Refer to lot expiration date on labels.
• Thawed clonal grade medium should be stored in the dark at 4°C for one month. Do not refreeze.
• Supplement may be thawed at room temperature, aliquoted and stored in the dark at 4°C for two months.
• After supplement is added to clonal grade medium, store in the dark at 4°C for up to
two weeks. Do not refreeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
ESGRO is a registered trademark of Merck KGaA, Darmstadt, Germany
cOmplete is a trademark of Roche

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic transcriptional events in embryonic stem cells mediated by the super elongation complex (SEC).
Lin C. et al.
Genes & Development null
HDAC1 regulates pluripotency and lineage specific transcriptional networks in embryonic and trophoblast stem cells.
Kidder B. L. & Palmer S.
Nucleic Acids Research null
A critical appraisal of factors affecting the accuracy of results obtained when using flow cytometry in stem cell investigations: where do you put your gates?
Hughes O. R. et al.
Cytometry. Part A : the Journal of the International Society For Analytical Cytology null
A potential use of embryonic stem cell medium for the in vitro culture of preimplantation embryos.
Gelber K. et al.
Journal of Assisted Reproduction and Genetics null
Optimized mouse ES cell culture system by suspension growth in a fully defined medium.
Andang M. et al.
Nature Protocols null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service