Skip to Content
Merck
All Photos(1)

Key Documents

SCT058

Sigma-Aldrich

BioTracker MB1 Tyrosinase Live Cell Dye

Live cell imaging dye for near-infrared visualization of cellular tyrosinase, which is overexpressed in melanoma.

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€342.00

€342.00


Estimated to ship on17 May 2025



Select a Size

Change View
1 MG
€342.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€342.00


Estimated to ship on17 May 2025


form

lyophilized

detection method

fluorometric

General description

Melanoma is a serious form of skin cancer that begins in cells known as melanocytes. Melanoma is an aggressive malignancy with rapid growth and early metastasis. Tyrosinase (TYR) is a potential biomarker for the detection of melanoma because its expression level is closely related with the level of malignancy. BioTracker MB1 Tyrosinase live cell dye is a near-infrared dye that selectively reacts with TYR but not with various potential interfering agents,including inorganic salts (Na+, K+, Ca2+, and Mg2+), aminoacids (Cys, Glu, and Arg), and other biological moleculesincluding reduced GSH, BSA, glucose, hydrogen peroxide,vitamin C, lysozyme, glucose oxidase (GOD), and TYR.Spectral Properties:Excitation: 640nmEmission: 706nm (Near-infrared)Research Category:Live Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service