Skip to Content
Merck
All Photos(1)

Key Documents

SCT050

Sigma-Aldrich

BioTracker DCM-Bgal Live Cell Dye

Near-Infrared live cell imaging dye for visualization of beta-Galactosidase activity, an important marker for cell senescence and primary ovarian cancers.

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€342.00

€342.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 MG
€342.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

€342.00


Estimated to ship on28 May 2025


form

lyophilized

detection method

fluorometric

General description

The BioTracker DCM-Bgal live cell dye is a ratiometric near-infrared (NIR) dye for the real-time fluorescent quantification of beta-galatosidase (ß-gal) enzyme activity in vitro, in vivo and in situ.ß-galactosidase is an important marker for cell senescence and primary ovarian cancers. BioTracker DCM-Bgal live cell dye has light-up ratiometric NIR fluorescence withlarge Stokes shift, high photostability than commercial ICG, and pH independency under the physiological range, allowing for the real-timeevaluation of β-gal activity. DCM-βgal displays higher affinity for β-galthan commercial X-gal and much faster response to β-gal thanthe previously reported FDG.Spectral Properties:NIR fluorescence images obtained from 605 to 725 nm, λex = 560 nmResearch Categories:Apoptosis & CancerCell ImagingOncology

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service