Skip to Content
Merck
All Photos(1)

Key Documents

SCT030

Sigma-Aldrich

BioTracker 575 Red Fe2+ Dye

Live cell imaging dye for iron (Fe2+) that localizes in golgi organelles.

Synonym(s):

Cellular iron detection, Live cell imaging probe, Live cell iron probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47
Pricing and availability is not currently available.

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Iron (Fe2+) is an essential element for blood production. About 70 percent of your body′s iron is found in the red blood cells of your blood called hemoglobin and in muscle cells called myoglobin. Iron is an essential metal for living organisms, but dysregulation of its homeostasis at the cellular level can trigger detrimental oxidative and/or nitrosative stress and damage events.

The BioTracker 575 Red Fe2+ Dye is a fluorescent imaging probe for the detection of iron (II) in living cells. This probe is localized in Golgi, and reacts selectively with Fe2+.

Spectral Properties
Absorbance: 540nm
Emission: 575nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Storage and Stability

Store BioTracker 575 Red Fe2+ Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service