Skip to Content
Merck
All Photos(1)

Key Documents

SCT140

Sigma-Aldrich

BioTracker 555 UV-Excitation Red Lysosome Dye

Live cell imaging dye for acidic cellular organelles such as lysosomes.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€301.00

€301.00


Estimated to ship on25 May 2025



Select a Size

Change View
50 μL
€301.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€301.00


Estimated to ship on25 May 2025


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Lysosomes are membrane-enclosed organelles that contain an array of enzymes capable of breaking down all types of biological material including proteins, nucleic acids, carbohydrates, and lipids. Lysosomes function as the digestive system of the cell, serving both to degrade material taken up from outside the cell and to digest obsolete components of the cell itself.

BioTracker Lysosome dyes are fluorescent stains for imaging lysosome localization and morphology in live cells. The dyes accumulate in the low pH environment of the lysosomes, resulting in highly specific lysosomal staining without the need for a wash step.

The BioTracker 555 UV-Excitation Red Lysosome is a red fluorogenic lysosome dye with pH-dependent fluorescence. The dye is unique among commercially available lysosome dyes in that its fluorescence in cells is activated by exposure to UV excitation. In solution, the dye shows pH-dependent fluorescence that does not require UV activation. The dye initially shows low fluorescence, but brief exposure to UV excitation from a mercury arc lamp through a DAPI filter.

Spectral Properties
Absorbance: 554nm
Emission: 583nm

Application

Live cell fluorescent imaging
Live cell imaging dye for acidic cellular organelles such as lysosomes.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Quality

Spectral Properties
Absorbance: 554nm
Emission: 583nm

Physical form

Liquid

Storage and Stability

Store BioTracker 555 UV-Excitation Red Lysosome Dye at -20ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service