Skip to Content
Merck
All Photos(1)

Key Documents

SCT028

Sigma-Aldrich

BioTracker 525 Green GGT Dye

Live cell imaging dye for Gamma-glutamyltransferase (GGT) for cancer research applications.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47
Pricing and availability is not currently available.

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Gamma-glutamyltransferase, or GGT, is an enzyme that plays a key role in the gamma-glutamyl cycle, a pathway for the synthesis and degradation of glutathione and drug and xenobiotic detoxification. GGT is predominantly used as a diagnostic marker for liver disease and is often significantly increased in human malignancies and cancers.

The BioTracker 525 Green GGT Dye is cancer-selective fluorescent imaging probe. This probe is rapidly activated by g-glutamyltranspeptidase (GGT), which is known to be tumor-associated enzyme, to give highly fluorescent compound. The membrane-permeable dye is applicable to live cell or in vivo imaging of cancer cells.

Spectral Properties
Absorbance: 496nm
Emission: 525nm

Storage and Stability

Store BioTracker 525 Green GGT Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service