Skip to Content
Merck

Skip To

SCC166

MS5-mDLL1 Stromal Cell Line

Mouse

Synonym(s):

MS-5-mDLL1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
eCl@ss:
32011203
Biological source:
mouse
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MS5-mDLL1 Stromal Cell Line, The MS5-mDLL1 stromal cell line is used in a serum-free artificial thymic organoid (ATO) system to generate mature T-cells from human hematopoietic stem and progenitor cells.

biological source

mouse

technique(s)

cell based assay: suitable, cell culture | mammalian: suitable

shipped in

ambient

storage temp.

-140 to -196°C

General description

In vivo, T-cells develop in the thymus from bone-marrow derived hematopoietic stem and progenitor cells (HSPCs). In vitro, various model systems have been employed to mimic thymic function supportive of T-cell development including the OP9-DL1 system. Recently, a serum-free artificial thymic organoid (ATO) system was developed that supports highly efficient and reproducible in vitro differentiation and positive selection of human T-cells from multiple sources of HSPCs, including cord blood, bone marrow and peripheral blood CD34+ HSPCs (Seet et al. 2017). A key component of the ATO system is the MS5-mDLL1 cell line which obviates the need for primary thymic tissues.
To generate the MS5-mDLL1, a murine stromal cell line (MS5) was transduced with a third-generation lentiviral vector encoding mouse DLL1 and mStrawberry.

To form organoids, MS5-mDLL1 stromal cells are aggregated with HSPCs by centrifugation and cultured on a cell culture insert at the air-fluid interface. Detailed protocols for setting up the artificial thymic organoid (ATO) system are described in reference (Seet et al. 2017).

Reference:
Seet, C.S., He, C., Bethune, M.T., Li, S., Chick, B., Gschweng, E.H., Zhu, Y., Kim, K., Kohn, D.B., Baltimore, D. and Crooks, G.M. (2017) Generation of mature T cells from human hematopoietic stem and progenitor cells in artificial thymic organoids. Nat Methods 14(5): 521-530.

Application

Research Category
Immune Response

Inflammation & Immunology
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.
The MS5-mDLL1 stromal cell line is used in a serum-free artificial thymic organoid (ATO) system to generate mature T-cells from human hematopoietic stem and progenitor cells.

Biochem/physiol Actions

Stromal Cells

Preparation Note

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Analysis Note

• Each vial contains ≥1X10⁶ viable cells.
• Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC167SCC136SCC065
description

The MS5-mDLL1 stromal cell line is used in a serum-free artificial thymic organoid (ATO) system to generate mature T-cells from human hematopoietic stem and progenitor cells.

description

The MS5-mDLL1 stromal cell line is used in a serum-free artificial thymic organoid (ATO) system to generate mature T-cells from human hematopoietic stem and progenitor cells.

description

-

description

HL-1 Cardiac Muscle Cell Line has been extensively characterized and is a valuable model system to address questions of cardiac biology at the cellular & molecular levels.

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

technique(s)

cell culture | mammalian: suitable, cell based assay: suitable

technique(s)

cell culture | mammalian: suitable, cell based assay: suitable

technique(s)

cell culture | mammalian: suitable, cell differentiation: suitable

technique(s)

cell culture | mammalian: suitable, cell based assay: suitable

shipped in

ambient

shipped in

ambient

shipped in

liquid nitrogen

shipped in

-

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

−196°C

storage temp.

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters