Skip to Content
Merck
All Photos(1)

Key Documents

SAE176-100UG

Sigma-Aldrich

SAvPhire Monomeric Streptavidin Alpha Fluor 488

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€372.00

€372.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€372.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€372.00


Check Cart for Availability

Request a Bulk Order

conjugate

horse radish peroxidase (HRP)

Quality Level

clone

monoclonal

species reactivity

all

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

General description

Upon reconstitution in 100 μL of distilled water the concentration will be 1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Specificity

SAvPhire Monomeric Streptavidin expressed in E. coli and conjugated with Alpha Fluor 488 that is suitable for immunodetection of biotinylated proteins.

Application

Epitope: Unknown

Quality

Evaluated by Immunocytochemistry in A431 cells.



Immunofluorescent analysis of A431 cells performed using a 1:1 mixture of biotinylated Anti-EGFR (1:1,000 dilution) and SAvPhire Monomeric Streptavidin Alpha Fluor 488 detected EGFR.

Physical properties

15.5 kDa for SAvPhire Monomeric Streptavidin (with an N-terminal His-tag and a C-terminal FLAG tag).

Physical form

SAvPhire Monomeric Streptavidin Alpha Fluor 488 lyophilized from PBS containing D-Mannitol and Sucrose without azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Predicted to react with all species.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service