Skip to Content
Merck
All Photos(1)

Key Documents

SAE0109

Sigma-Aldrich

SAvPhire Monomeric Streptavidin alkaline phosphatase conjugate

Synonym(s):

SAVphire-AP, mSA-AP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

Escherichia coli

conjugate

alkaline phosphatase

clone

monoclonal

mol wt

15.5  kDa

technique(s)

immunohistochemistry: suitable (Paraffin)

General description

for Savphire monomeric streptavidin with an N-terminal His-tag and a C-terminal FLAG® tag.

Specificity

SAvPhire Monomeric Streptavidin expressed in E. coli and conjugated with alkaline phosphatase is suitable for immunodetection applications. SAvPhire Monomeric Streptavidin is expressed with an N-terminal His-tag and a C-terminal FLAG® tag.

Species Reactivity Note: Predicted to react with all species.

Immunogen

N/A

Application

SAvPhire Monomeric Streptavidin,Cat.#SAE0109, is a high-affinity recombinant variant of SA that minimizes the risk of aggregation, precipitation, and crosslinking in pre-incubation with biotinylated molecules and offers easier access to the desired targets, tested for use in IHC (Paraffin).

Quality

Evaluated by Immunohistochemistry (Paraffin) with Anti-NeuN antibody (ABN78) for detection of NeuN in mouse brain tissue sections.

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:500 dilution Anti-NeuN (ABN78) was used to stain sections of mouse cerebral cortex and signal was detected with Savphire monomeric streptavidin alkaline phosphatase conjugate (1:500 dilution) and Vulcan Fast Red.

Physical form

Monomeric Streptavidin in buffer containing 0.05 M Tris, 0.15 M NaCl, 1 mM MgCl2, 0.2% BSA, pH 8.0 with 0.1% sodium azide with 10% glycerol.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service