Skip to Content
Merck
All Photos(1)

Key Documents

PX1834

Supelco

2-Propanol

≥99.9% (GC), suitable for LC/MS, gas chromatography (GC), HPLC, suitable for SM 4500 - NH3, suitable for SM 2320, suitable for SM 2310, OmniSolv®

Synonym(s):

2-Propanol, 2-Propanol,IPA, 2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-661-7
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

Product Name

Isopropyl Alcohol, OmniSolv®

Agency

suitable for SM 2310
suitable for SM 2320
suitable for SM 4500 - NH3

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
43 hPa ( 20 °C)
44 mmHg ( 25 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O)
≤0.2 μeq/g Titrable acid
≤2 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

UV cutoff

≤204 nm

UV absorption

λ: 204 nm Amax: ≤1.00
λ: 205 nm Amax: ≤0.80
λ: 210 nm Amax: ≤0.35
λ: 220 nm Amax: ≤0.10
λ: 230 nm Amax: ≤0.05
λ: 240 nm Amax: ≤0.02
λ: 260 nm Amax: ≤0.005
λ: 300 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®
PX1834P-1 is packaged in poly coated glass bottles

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minCapillary ECD responsive substances (as PCNB)2ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidIdentity (IR-spectrum)ConformsRefractive index (at 20°C)Titrable acid0.2µeq/g maxUV Abs. at 204 nm1.00AU maxUV Abs. at 205 nm0.80AU maxUV Abs. at 210 nm0.35AU maxUV Abs. at 220 nm0.10AU maxUV Abs. at 230 nm0.05AU maxUV Abs. at 240 nm0.02AU maxUV Abs. at 260 nm0.005AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off204nm maxWater (H2O)0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service