AX0441
Reagent Alcohol
94.0-96.0%, 89.0-92.0 % (v/v) ethanol, Reagent, Meets ACS Specifications GR ACS
About This Item
Recommended Products
Product Name
Alcohol, Reagent, Meets ACS Specifications GR ACS
grade
reagent
Quality Level
200
300
Assay
94.0-96.0%
form
liquid
composition
methanol, 3.5-5.5%
iso-propyl alcohol, 4.0-6.0%
concentration
89.0-92.0 % (v/v) ethanol
dilution
(for analytical testing)
impurities
≤0.075% Water (H2O)
evapn. residue
≤0.001%
color
clear colorless
APHA: ≤10
transition temp
flash point 12 °C
shipped in
ambient
storage temp.
room temp
SMILES string
OCC
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Prior to shipment of unit sizes of Reagent Alcohol over 4L, a letterhead application approved by the Bureau of Alcohol, Tobacco & Firearms (ATF) must be on file with EMD Chemicals.
Used as the final stage in dehydration for processing tissue especially in electron microscopy.
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Physical form
Analysis Note
Assay (ethanol): 89.0-92.0% (v/v)
Assay (isopropyl alcohol): 4.0-6.0% (v/v)
Assay (methanol): 3.5-5.5% (v/v)
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.075% max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
53.6 °F - closed cup
Flash Point(C)
12 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service