PCC030C05C
Pellicon® Capsule with Ultracel® Membrane
pore size 30 kDa, C Screen, size 0.5 m2 , with AseptiQuik® G connectors
Select a Size
+43 1 3602 77 47 43or
This item is not able to be purchased as a guest user. For further information, please contact us at .Select a Size
About This Item
+43 1 3602 77 47 43or
This item is not able to be purchased as a guest user. For further information, please contact us at .Recommended Products
material
EPDM rubber seal
PPO/PS blend (core)
PPO/PS blend housing (PPO/PS Blend)
TPE seal
Ultracel® membrane
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene screen
polyurethane adhesive
Quality Level
sterility
sterile; γ-irradiated
product line
Pellicon®
storage condition
room temperature (15-30°C)
parameter
4-8 L/min-m2 flow rate (Feed)
50 psi max. transmembrane pressure (3.5 bar) at 4-30 °C (Forward)
75 psi max. inlet pressure (5.1 bar) at 4-30 °C
technique(s)
ultrafiltration: suitable
L
41.1 cm (16.2 in.)
diam.
5.7 cm (2.3 in.)
filtration area
0.5 m2
screen size
, Type C screen (coarse screen)
size
0.5 m2 , with AseptiQuik® G connectors
volume
119 mL , in feed channel
149 mL , in permeate channel
impurities
≤5 ppm Meets TOC (in retentate effluent, after a WFI flush of 20 L/m2)
matrix
Ultracel®
pore size
30 kDa
pH range
2-13
fitting
inlet/outlet connection (Feed: AseptiQuik® G Connector)
inlet/outlet connection (Permeate: AseptiQuik® G Connector)
inlet/outlet connection (Retentate: AseptiQuik® G Connector)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
Gamma sterilized and preseravtive-free - ready to process in minutes
True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families
Packaging
Other Notes
Comprehensive documentation to support qualification, risk assessment and process optimization needs.
Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
Convenient 24/7 access to up-to-date product information.
Legal Information
related product
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Find best practices for using Pellicon® Capsules in ADC processing as well as data from two studies using Pellicon® Capsules and Pellicon® cassettes in the ADC manufacturing process.
Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.
Related Content
Learn more on mAb downstream processing, more specifically concentration and diafiltration, and the relevant associated products.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service