Skip to Content
Merck

Skip To

P3C030D05

Pellicon® 3 Cassette with Ultracel® Membrane

filtration area 0.57 m2, NMWCO 30 kDa

Synonym(s):

Pellicon® 3 Cassette with Ultracel 30 kDa Membrane, D screen, 0.57 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32031602
UNSPSC Code:
41104903
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

TPE seal, composite regenerated cellulose (CRC) , polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

feature

holdup volume 118 mL (in feed channel)

manufacturer/tradename

Pellicon®

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 40 psi max. transmembrane pressure at 4-50 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C

technique(s)

analytical sample preparation: suitable, buffer exchange: suitable, protein purification: suitable (concentration)

H

2.6 cm (1.01 in.)

L

20.6 cm (8.1 in.)

W

17.8 cm (7 in.)

filtration area

0.57 m2

membrane area

0.57 m2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel® (Membrane)

pore size

30 kDa NMWCO

pH

2-13

General description

Pellicon® 3 filtration devices are advanced, high-performance cassettes ideal for today’s higher titer therapeutic antibodies as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.

The Pellicon® 3 Cassette with Ultracel® Membrane is designed for the filtration of therapeutic proteins (such as monoclonal antibodies) and recombinant and non-recombinant proteins.

This P3C030D05 Pellicon® 3 ultrafiltration device features a pore size of 30 kDa NMWCO, a filtration area of 0.57 m2, a PLCTK membrane, and dimensions measuring 2.4 cm (H) x 5.6 cm (W) x 20.6 cm (L).

Features and Benefits

  • Rugged & reliable – ideally suited for today′s more demanding filtration processes
  • High performance – produces excellent resolution, yields and back-pressure resistance
  • Optimum product recovery – thanks to proven membrane technology
  • Void-free technology – leading-edge membranes resolve virtually any separation challenge
  • Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
  • Automated manufacturing – delivers unbeatable performance, consistency and reliability
  • Easy installation & care – simple to install and maintain

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P3C005C05P3C010C05P3C030C01
pore size

30 kDa NMWCO

pore size

5 kDa NMWCO

pore size

10 kDa NMWCO

pore size

30 kDa NMWCO

technique(s)

analytical sample preparation: suitable, protein purification: suitable (concentration), buffer exchange: suitable

technique(s)

analytical sample preparation: suitable, buffer exchange: suitable, protein purification: suitable (concentration)

technique(s)

analytical sample preparation: suitable, buffer exchange: suitable, protein purification: suitable (concentration)

technique(s)

analytical sample preparation: suitable, buffer exchange: suitable, protein purification: suitable (concentration)

filtration area

0.57 m2

filtration area

0.57 m2

filtration area

0.57 m2

filtration area

0.11 m2

matrix

Ultracel® (Membrane)

matrix

Ultracel® (Membrane)

matrix

Ultracel® (Membrane)

matrix

Ultracel® (Membrane)

material

TPE seal, polypropylene screen, composite regenerated cellulose (CRC) , stainless steel cap (for end; polypropylene-encapsulated)

material

TPE seal, composite regenerated cellulose (CRC) , polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

material

TPE seal, composite regenerated cellulose (CRC) , polypropylene screen, stainless steel cap (for end; polypropylene-encapsulated)

material

TPE seal, composite regenerated cellulose (CRC) , polypropylene cap (for end), polypropylene screen

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 40 psi max. transmembrane pressure at 4-50 °C, 80 psi max. transmembrane pressure at 4-40 °C, 4-6 L/min-m2 recirculation rate, 6.9 bar max. inlet pressure (100 psi)

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 40 psi max. transmembrane pressure at 4-50 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 40 psi max. transmembrane pressure at 4-50 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 40 psi max. transmembrane pressure at 4-50 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters