Skip to Content
Merck
All Photos(6)

Key Documents

MMSMCRA48

Millipore

Milliflex Oasis® Culture Media Plate

Milliflex Oasis® Culture Media Plate

suitable for (R2A low nutrient media plates for waterborne microorganisms)

Synonym(s):

Blue coded, ready-to-use agar plate for use with Milliflex Oasis® system, MILLIFLEX OASIS® R2A low nutrient media plates

Sign Into View Organizational & Contract Pricing

Select a Size

48 EACH
€292.00

€292.00


Estimated to ship on12 April 2025


Request more information

Select a Size

Change View
48 EACH
€292.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

€292.00


Estimated to ship on12 April 2025


Request more information

material

blue polystyrene plate

Quality Level

Agency

(EP, USP, JP)

sterility

aseptically filled

form

plate (Pre-poured agar)

shelf life

12 mo.

feature

ready-to-use

packaging

pk of 48 tests

manufacturer/tradename

Milliflex Oasis®

pH

7.2±0.2 (25 °C)

suitability

suitable for (R2A low nutrient media plates for waterborne microorganisms)
suitable for (suitable for solid cultivation)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with MILLIFLEX OASIS® system

storage temp.

2-25°C

Looking for similar products? Visit Product Comparison Guide

General description

MILLIFLEX OASIS® R2A low nutrient media plates are ready-to-use agar-based media for convenient, rapid and reliable testing. These unique blue colour coded plates offer the control of critical handling steps all along the workflow in conjugation with the MILLIFLEX OASIS® system. They are manufactured and QC tested according to pharmaceutical industry requirements. The plates can be stacked and locked to ensure safe transportation and avoid accidental opening. The empty funnel tray can also be used as an incubation rack.

MILLIFLEX® TRACE features ensures total traceability through unique 2D barcodes identification and detachable labels. If left unopened, the plates remain sealed unlike standard petri plates which reduces condensation and dehydration for an extended storage time owing to its long shelf life.

Application

MILLIFLEX OASIS® R2A low nutrient media plates are used for the recovery of stressed heterotrophic plate count bacteria found in various types of waters, especially high purity water, Beer and Bottled Water. They are widely used in pharmaceutical and cosmetic industry.

Physical form

48 ready-to-use pre-poured agar plates

Analysis Note

The Colonies appear clear to creamy white, some may produce pigment

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex Oasis® system monitors bioburden in various samples, increasing lab productivity.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service