Skip to Content
Merck
All Photos(5)

Key Documents

MMHEADMM1

Millipore

Milliflex Oasis® Filtration Head

for use with Milliflex Oasis® filtration units, designed for MMSYSTMM1 pump

Synonym(s):

Filtration Accessory, Filtration Head, MILLIFLEX OASIS® filtration head, Milliflex Oasis® filtration pump head

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€506.00

€506.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 UNIT
€506.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

€506.00


Check Cart for Availability

Request more information

material

316L stainless steel (ASI)

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® filtration units, designed for MMSYSTMM1 pump

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Milliflex Oasis® filtration system offers a solution to simplify and secure the microbial testing process with premium level productivity, result reliability and regulatory compliance. The Milliflex Oasis® filtration head is made of 316L stainless steel and can be autoclaved at 121°C for up to 30 minutes or at 134°C for up to 15 minutes. The Milliflex Oasis® filtration head has been designed to fit the Milliflex Oasis® vacuum pump MMSYSTMM1.

Application

Milliflex Oasis® FiltrationHead is used for the filtration of liquid samples for Bioburden testing, watermonitoring, and general microbiological analysis of various filtrable fluids.

Other Notes

In the European Union, this product will be considered Waste Electric and Electronic Equipment (WEEE) at the end of its life. Please click here for details on returning it for recycling.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service