Skip to Content
Merck
All Photos(3)

Key Documents

MIHABG250

Millipore

Microfil® funnel with S-Pak®

sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black filter, gridded filter

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

black filter
gridded filter
mixed cellulose esters (MCE)
polypropylene

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic

manufacturer/tradename

Microfil® with S-Pak®

parameter

250 mL sample volume

technique(s)

sterile filtration: suitable

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.45 μm pore size

input

sample type beverage(s)
sample(s) (environmental)
sample(s) (environmental)
sample type food(s)

suitability

suitable for microbiology

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Microfil® with S-Pak® filters assure leak-free operation for uniform microorganism recoveries and an easy transfer of the membrane with forceps, preventing accidental touching of the filtration area. Combine the S-Pak® system with the Microfil® filtration unit for the ultimate convenience and efficiency. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use 250 mL Microfil® funnels that come with the 0.45 μm pore size, black gridded S-Pak® filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient for general microbiological analysis. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in uniform recovery of organisms.

Application

Microfil® with S-Pak® filters find application in the Clinical, Cosmetics, Environmental, Industrial QC, Pharmaceutical, Food, and Beverage industries. Microbial analysis of beer, bottled water, cider, sports drink, and wine by membrane filtration method

Features and Benefits

  • Convenient and easy to use
  • Sterilized, saves time and expense
  • Reduce laboratory workload
  • Faster filtration rate
  • Consistent recoveries

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service