Skip to Content
Merck
All Photos(5)

Key Documents

MCFLX4710

Millipore

MILLICUP-FLEX, 47 mm Disposable Filtration Unit

250 mL, Solvent-Resistant, GL45 Connection, Single-Use, Nonsterile

Synonym(s):

Disposable filtration unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101708
NACRES:
NB.43

material

polypropylene device

sterility

non-sterile

technique(s)

vacuum filtration: suitable

funnel capacity

250 mL

fitting

inlet funnel
outlet bottle top

application(s)

filtration

General description

The Millicup-FLEX nonsterile, vacuum-driven filtration unit combines the convenience of disposable filtration with the flexibility and compatibility of traditional vacuum filtration glassware. With solvent-resistant, modular components, the Millicup-FLEX filtration unit offers a safe and efficient means for filtering a variety of aqueous and organic solutions, allowing the user to filter directly into GL45 vacuum-rated filtration bottles with their choice of 47mm disc membranes (sold separately). The ready-to-assemble, clampless design of the Millicup-FLEX filtration system eliminates the need for cleaning prior to filtration – saving time and ultimately reducing the risk of sample contamination. Following filtration, the 250mL funnel and membrane support collar easily disconnect to enable direct access to the membrane.

Features & Benefits:
  • Compatible with organic and aqueous solvents
  • Ergonomic, clampless design
  • Reduce contamination risk
  • Filter directly into vacuum-rated storage bottles
  • Easy access to membrane after filtration (membranes sold separately)
  • Fully recyclable components


Available in 2 versions:
  • Semi-reusable Kit: 10 funnels with 2 semi-reusable membrane support collars and vacuum tube adapters (MCFLX4702)
  • Single-Use Kit: 10 funnels with 10 single-use membrane support collars and vacuum tube adapters (MCFLX4710)


All three components of the Millicup-FLEX filtration unit are fully recyclable, minimizing your environmental impact.

Application

  • Laboratory Filtration
  • Air/gas/organic solvent filtration and venting
  • Clarification of water, buffers or aqueous solutions
  • Environmental Analysis
  • Food and Beverage
  • Industrial QC
  • Pharmaceutical Products Analysis
  • Water Monitoring

Direct patient care: No

Components

Kit includes: 10 funnels, 10 support collars, and 10 vacuum tube adapters

Principle

Vacuum

Physical form

Color code: Clear with blue adapter

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Methods and tutorial video for filtration techniques in contamination analysis using a faster alternative to glass vacuum filtration systems.

Learn how filter selection and sample and mobile phase filtration impact HPLC back pressure and column lifetime.

Data and recommended filtration products for OTM-45 Method for Air Monitoring of PFAS Compounds.

Data and recommended filtration products for PFAS testing using Modified EPA 1633.

See All

Protocols

Millicup™-FLEX disposable vacuum filter was designed to reduce contamination and save time. The solvent-resistant unit allows for filtration into storage bottles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service