EFHAB10MS
EZ-Fit® filtration unit without absorbent pad
0.45 µm MCE black membrane with 100 mL funnel, multi packaging, suitable for solid cultivation
Synonym(s):
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
About This Item
Recommended Products
material
black filter
gridded filter
mixed cellulose esters (MCE)
styrene-butadiene device
Quality Level
Agency
ISO 7004:1985
sterility
sterile; electron beam irradiated
manufacturer/tradename
EZ-Fit®
packaging
pkg of Multi packaging
parameter
100 mL sample volume
filter diam.
47 mm
funnel capacity
100 mL
pore size
0.45 μm pore size
input
sample type food(s)
sample type pharmaceutical(s)
sample(s) (environmental)
sample type beverage(s)
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.
The EZ-FIT® Azure feature makes the EZ-FIT® filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.
The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT® Manifold, which features specific EZ-FIT® Filtration heads.
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service