Skip to Content
Merck
All Photos(2)

Key Documents

MABT1488

Sigma-Aldrich

Anti-MBP (Citrulline R130) Antibody, clone 3C6

clone 3C6, from mouse

Synonym(s):

Myelin basic protein, Myelin A1 protein, Myelin membrane encephalitogenic protein, Citrullinated MBP

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€129.00
100 μG
€353.00

€129.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€129.00
100 μG
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€129.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C6, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

citrullination (Arg130)

General description

Myelin basic protein (UniProt: P02686; also known as MBP, Myelin A1 protein, Myelin membrane encephalitogenic protein) is encoded by the MBP gene (Gene ID: 4155) in human. MBP is a homodimeric protein that is found in both the central and the peripheral nervous system. It is one of the most abundant protein component of the myelin membrane in the CNS and plays a role in both the formation and stabilization of this myelin membrane. It is responsible for adhesion of the cytosolic surfaces of multilayered compact myelin. MBP can interact with many polyanionic proteins including actin, tubulin, calmodulin, and clathrin, and with negatively charged lipids. MBP isoforms are found in both the central and the peripheral nervous system, whereas Golli-MBP isoforms are expressed in fetal thymus, spleen and spinal cord, as well as in cell lines derived from the immune system. MBP expression begins abruptly in 14-16-week-old fetuses. Even smaller isoforms seem to be produced during embryogenesis; some of these persisting in the adult. MBP citrullination by peptidylarginine deiminase (PAD) leads to incomplete protein-lipid bilayer interactions and increases its vulnerability to proteolytic enzymes, resulting in disorganization of the myelin sheath in the central nervous system. Citrullinated MBP (citMBP) has been suggested to be a hallmark of demyelination. MBP citrullination has frequently been demonstrated in various CNS disorders. citMBP at R25, R122, and R130 is reported in the brains of subjects with Creutzfeldt-Jakob disease (CJD). Clone 3C6 specifically recognizes citMBP at R130. (Ref.: Jang, B., et al. (2017). Mol. Neurobiol. 55(4); 3172-3184; Boggs, JM (2006). Cell. Mol. Life Sci. 63(17); 1945-1961).

Specificity

Clone 3C6 is a mouse monoclonal antibody that detects human and murine myelin basic protein citrullinated on residue 130. It targets an epitope with in 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from isoform 5 of human Myelin Basic Protein with citrullination on residue 130.

Application

Anti-MBP (Citrulline R130), clone 3C6, Cat. No. MABT1488, is a mouse monoclonal antibody that detects Myelin basic protein citrullinated on residue 130 and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected MBP (Citrulline R130) in Immunohistochemistry applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MBP (Citrulline R130) in Myelin basic protein (MBP) treated with human PAD2 (Courtesy of Byungki Jang, Ph.D./Eun-Kyoung Choi, Ph.D., Ilsong Institute of Life Science, Hallym University, South Korea).

Western Blotting Analysis: A representative lot detected MBP (Citrulline R130) in Western Blotting applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).

ELISA Analysis: A representative lot detected MBP (Citrulline R130) in ELISA applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in GST-tagged myelin basic protein (MBP) treated with human PADI-2.

Western Blotting Analysis: 4 µg/mL of this antibody detected MBP (Citrulline R130) in GST-tagged myelin basic protein (MBP) treated with human Peptidyl Arginine Deiminase 2 (PAD-2)(200 ng for 4 h at 37°C).

Target description

~41 kDa observed; 18.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service