MABT1510
Anti-MBP (Citrulline R25) Antibody, clone 1B8
clone 1B8, from mouse
Synonym(s):
Myelin basic protein, Myelin A1 protein, Myelin membrane encephalitogenic protein, Citrullinated MBP
Select a Size
€152.00
Estimated to ship on29 May 2025
Select a Size
About This Item
€152.00
Estimated to ship on29 May 2025
Recommended Products
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
1B8, monoclonal
species reactivity
human, mouse
packaging
antibody small pack of 25 μg
technique(s)
ELISA: suitable
electron microscopy: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
target post-translational modification
citrullination (Arg25)
General description
Specificity
Immunogen
Application
Electron Microscopy Analysis: A representative lot detected MBP (Citrulline R25) in Electron Microscopy applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MBP (Citrulline R25) in Myelin basic protein (MBP) treated with human PAD2 (Courtesy of Byungki Jang, Ph.D./Eun-Kyoung Choi, Ph.D., Ilsong Institute of Life Science, Hallym University, South Korea).
Immunofluorescence Analysis: A representative lot detected MBP (Citrulline R25) in Immunofluorescence applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).
Western Blotting Analysis: A representative lot detected MBP (Citrulline R25) in Western Blotting applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).
ELISA Analysis: A representative lot detected MBP (Citrulline R25) in ELISA applications (Jang, B., et. al. (2018). Mol Neurobiol. 55(4):3172-3184).
Cell Structure
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected Myelin basic protein (MBP) treated with human human Peptidyl Arginine Deiminase 2 (PAD-2) (200 ng for 4 hours at 37°C) with R25 citrullination.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service