Skip to Content
Merck
All Photos(2)

Key Documents

MABS2068

Sigma-Aldrich

Anti-HSD3B1 Antibody, clone 4C1

clone 4C1, from mouse

Synonym(s):

3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3-beta-HSD I, Trophoblast antigen FDO161G

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€315.00

€116.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€315.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... HSD3B1(3283)

General description

3 beta-hydroxysteroid dehydrogenase/Delta 5→4-isomerase type 1 (UniProt: P14060; also known as 3 beta-hydroxysteroid dehydrogenase/Delta 5→4-isomerase type I, 3-beta-HSD I, Trophoblast antigen FDO161G) is encoded by the HSD3B1 (also known as 3BH, HSDB3A) gene (Gene ID: 3283) in human. HSD3B1 is a single -pass membrane protein localized in mitochondria and endoplasmic reticulum. Its transmembrane region is localized to amino acids 288-308. It is a bifunctional enzyme that catalyzes the oxidative conversion of Delta 5-ene-3-beta-hydroxy steroid and causes oxidative conversion of ketosteroids. It plays a vital role in the biosynthesis of all classes of hormonal steroids. Humans have two HSD3B isozymes. HSD3B1 is mainly expressed in the placenta, mammary and, prostate glands, and other peripheral tissues including the skin and adipose tissue and HSD3B2 is expressed primarily in the adrenal and gonads. They exhibit about 93% sequence homology. (Ref.: Gomez-Sanchez, CE et al. (2017). Steroids 127; 56-61).

Specificity

Clone 4C1 detects 3-beta-HSD I (HSD3B1) in human tissues. It targets an epitope with in 14 amino acids from the N-terminal region.

Immunogen

A KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human 3 beta-hydroxysteroid dehydrogenase/Delta 5→4-isomerase type 1 (HSD3B1).
Epitope: N-terminus

Application

Anti-HSD3B1, clone 4C1, Cat. No. MABS2068, is a mouse monoclonal antibody that detects 3-beta-HSD I (HSD3B1) and has been tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected HSD3B1 in human adrenal gland tissue sections.

ELISA Analysis : A representative lot detected HSD3B1 in ELISA applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).

Western Blotting Analysis: 1 µg/mL from a representative lot detected HSD3B1 in human placenta tissue lysate in Western blotting applications.

Immunohistochemistry (Paraffin) Analysis: A representative lot detected HSD3B1 in Immunohistochemistry applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).

Western Blotting Analysis: A representative lot detected HSD3B1 in Western Blotting applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).
Research Category
Signaling

Quality

Evaluated by Immunohistochemistry (Paraffin) in human placenta tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected HSD3B1 in human placenta tissue sections.

Target description

~38 kDa observed; 42.25 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service