Skip to Content
Merck
All Photos(1)

Key Documents

MABS2049

Sigma-Aldrich

Anti-Adenylate Cyclase 5 Antibody, clone 19D5.C1

clone 19D5.C1, from mouse

Synonym(s):

Adenylate cyclase type 5, EC: 4.6.1.1, ATP pyrophosphate-lyase 5, Adenylate cyclase type V, Adenylyl cyclase 5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€403.00

€154.00


Estimated to ship on24 May 2025



Select a Size

Change View
25 μG
€154.00
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on24 May 2025


biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

19D5.C1, monoclonal

species reactivity

rat, porcine, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Adcy5(224129)

General description

Adenylate cyclase type 5 (P84309; also known as EC: 4.6.1.1, ATP pyrophosphate-lyase 5, Adenylate cyclase type V, Adenylyl cyclase 5, AC5) is encoded by the Adcy5 gene (Gene ID: 224129) in murine species. Adenylate cyclase catalyzes the formation of cyclic AMP in response to G-protein signaling. It regulates the increase of free cytosolic calcium in response to increased blood glucose levels and contributes to the regulation of calcium-dependent insulin secretion. Adenylyl cyclases are typically made up of two clusters of six transmembrane domains separating two intracellular, overlapping catalytic domains. The amino acid sequence of each cytoplasmic domain that contains the nucleotide (ATP)-binding site, is well conserved among various subtypes. Thus far 9 different hormone-sensitive, membrane-bound AC isoforms have been identified. Adenylyl Cyclase 5 deficiency is shown to protect against diet-induced obesity and insulin resistance in mice and extends their life span. Adenylate cyclase 5 knockout mice display signs of reduced stress and protection against the stress of chronic pressure overload on heart. Adenylate cyclase 5 activity is stimulated by forskolin and is inhibited by adenosine and ATP analogs. (Ref.: Ho, D., et al. (2015). Diabetes. 64(7); 2636-2645. Vatner, SF., et al. (2015). Oxid. Med. Cell. Longev. 2015:250310).

Specificity

Clone 19D5.C1 detects Adnylate Cyclase 5 in murine, rat, and procine cells. It targets an epitope within 20 amino acids from the cytoplasmic domain.

Immunogen

Epitope: cytoplasmic domain
KLH-conjugated linear peptide corresponding to 20 amino acids from the cytoplasmic domain of murine adenylate cyclase V.

Application

Anti-Adenylate Cyclase 5, clone 19D5.C1, Cat. No. MABS2049, is a mouse monoclonal antibody that detects adenylate cyclase 5 and has been tested for use in ELISA and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected Adenylyl Cyclase 5 in Western Blotting applications (Hu, C.L., et. al. (2009). Am J Physiol Heart Circ Physiol. 297(5):H1776-82; De Lorenzo, M.S., et. al. (2014). Aging Cell. 13(1):102-10).

ELISA Analysis: A representative lot detected Adenylyl Cyclase 5 in ELISA applications (Hu, C.L., et. al. (2009). Am J Physiol Heart Circ Physiol. 297(5):H1776-82).

Quality

Evaluated by Western Blotting in mouse brain membrane preparations.

Western Blotting Analysis: 1 µg/mL of this antibody detected Adenylate Cyclase 5 in mouse brain membrane preparations.

Target description

~150 kDa observed; 139.12 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service