Skip to Content
Merck
All Photos(1)

Key Documents

MABF2086

Sigma-Aldrich

Anti-Adenylate Cyclase Toxin Antibody, B. pertussis Antibody, clone 3D1

clone 3D1, from mouse

Synonym(s):

Adenylate cyclase toxin, Bordetella pertussis Toxin, ACT, CyaA

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€403.00

€154.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€154.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D1, monoclonal

species reactivity

bacteria

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
inhibition assay: suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

target post-translational modification

unmodified

General description

Adenylate Cyclase Toxin (UniProt: C8C508; also known as ACT or CyaA) is one of several virulence factors produced by the bacterium Bordetella pertussis. Bordetella pertussis, a gram-negative bacterium, is shown to be the causative agent for whooping cough. It produces several virulence factors, including Cya A. The CyaA toxin is a bifunctional protein of 1,706 residues consisting of an NH2-terminal catalytic domain of 400 amino acids and a COOH-terminal part with 1,306 residues. The CyaA polypeptide is synthesized as an inactive protoxin, which is then converted to an active toxin by posttranslational palmitoylation of two internal lysine residues (Lys 856 and 963). Upon interaction of its C-terminal hemolysin moiety with the cell surface receptor alphaM-beta2 integrin ((Cd11b/Cd18), the N-terminal cyclase domain translocates into the host cell cytosol where it generates supraphysiological levels of cAMP concentration, which inhibits host cell anti-bacterial activities and apoptotic death of macrophages. CyaA toxin is also shown to elicit potassium ion efflux from erythrocytes in a process that is thought to be a precursor event to osmotic lysis of erythrocytes.Clone 3D1 is shown to cause both an enhancement of hemolysis and inhibition of intoxication in erythrocytes. It also inhibits toxin-induced cAMP accumulation in Jurkat cells. (Ref.: Guermonprez, P., et al. (2001). J. Exp. Med. 193(9); 1035 1044; Gray, MC., et al. (2001). J. Bacteriol. 183(20); 5904-5910; Holubova, J., et al. (2012). Infect. Immun. 80(3); 1181 1192).

Specificity

Clone 3D1 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis.

Immunogen

Adenylate cyclase (AC) toxin from Bordetella pertussis.

Application

Anti-Adenylate Cyclase Toxin, B. pertussis, clone 3D1 , Cat. No. MABF2086, is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis and has been tested for use in ELISA, Flow Cytometry, Inhibition Assay, Neutralizing, and Western Blotting.
Inhibition Analysis: A representative lot inhibited AC toxin-induced cAMP accumulation in Jurkat cells and inhibited intoxication of sheep erythrocytes. (Bumba, L., et. al. (2016). Mol Cell. 62(1):47-62; Gray, M.C., et. al. (2001). J Bacteriol. 183(20):5904-10; Holubova, J., et. al. (2012). Infect Immun. 80(3):1181-92; Hewlett, E.L., et. al. (2006). Mol Microbiol. 59(2):447-59).

Flow Cytometry Analysis: A representative lot detected Adenylate Cyclase Toxin, B. pertussis in Flow Cytometry applications (Gray, M.C., et. al. (2001). J Bacteriol. 183(20):5904-10).

Western Blotting Analysis: A representative lot detected Adenylate Cyclase Toxin, B. pertussis in Western Blotting applications (Lee, S.J., et. al. (1999). Infect Immun. 67(5):2090-5).

ELISA Analysis: A representative lot detected Adenylate Cyclase Toxin, B. pertussis in ELISA applications (Weingart, C.L., et. al. (2000). Infect Immun. 68(12):7152-5; Wang, X., et. al. (2015). J Biol Chem. 290(38):23025).

Neutralizing Analysis: A representative lot neutralized Adenylate Cyclase Toxin action. (Holubova, J., et. al. (2012). Infect Immun. 80(3):1181-92.

Quality

Evaluated by Western Blotting in Adenylate cyclase toxin from Bordetella pertussis.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Adenylate Cyclase Toxin from Bordetella pertussis.

Target description

~220 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service