Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
11H4, monoclonal
species reactivity
rat, mouse, human
packaging
antibody small pack of 25 μg
technique(s)
immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... LRP1(4035)
General description
Immunogen
Application
Immunoprecipitation Analysis: A representative lot detected LRP-1 in Immunoprecipitation applications (Nieves, E.C., et. al. (2010). J Biol Chem. 285(17):12595-603).
Western Blotting Analysis: A representative lot detected LRP-1 in Western Blotting applications (Spoelgen, R., et. al. (2009). Neuroscience. 158(4):1460-8; Lleó, A., et. al. (2005). J Biol Chem. 280(29):27303-9; Campana, W.M., et. al. (2006). J Nuerosci. 26(43):11197-207).
Immunocytochemistry Analysis: A representative lot detected LRP-1 in Immunocytochemistry applications (Lleó, A., et. al. (2005). J Biol Chem. 280(29):27303-9).
Immunohistochemistry Analysis: A representative lot detected LRP-1 in Immunohistochemistry applications (Campana, W.M., et. al. (2006). J Nuerosci. 26(43):11197-207).
Signaling
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected LRP-1 in human brain tissue.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified antibody | antibody form serum | antibody form purified antibody | antibody form purified immunoglobulin |
| clone 11H4, monoclonal | clone polyclonal | clone 8G1, monoclonal | clone LRP1-11, monoclonal |
| biological source mouse | biological source rabbit | biological source mouse | biological source mouse |
| species reactivity rat, mouse, human | species reactivity mouse | species reactivity human | species reactivity human, rat, mouse |
| Gene Information human ... LRP1(4035) | Gene Information human ... LRP1(4035) | Gene Information - | Gene Information human ... LRP1(4035) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



