Skip to Content
Merck
All Photos(1)

Key Documents

MABN224

Sigma-Aldrich

Anti-Thorase Antibody, clone 9G7.1

clone 9G7.1, from mouse

Synonym(s):

ATPase family AAA domain-containing protein 1, Thorase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on05 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9G7.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATAD1(84896)

General description

Thorase is an ATPase that plays a critical role in regulating the surface expression of AMPA receptors (AMPAR). Thorase thus helps mediate synaptic plasticity and learning and memory. Thorase expression is required for NMDA-stimulated AMPAR internalization and for the inhibition of GRIA1 and GRIA2 recycling back to the plasma membrane as both of these require ATP processing by Thorase. Thorase is vital for keeping in balance the molecular construction-deconstruction cycle that is required for memory formation. Thorase is one of many AAA+ ATPases that drive the assembly of proteins needed to form specialized receptors at the surfaces of synapses. These receptors are stimulated by neighboring neurons, setting up the signaling and answering connections vital to brain function. Thorase regulates the all-important complementary process of receptor disassembly at synapses, which ultimately tamps down signaling. Thorase is a postsynaptic localized protein, and also is associated with the peroxisome and cell junctions and synapses. Thorase is expressed in the brain and testis primarily.

Immunogen

GST-tagged recombinant protein corresponding to human Thorase.

Application

This Anti-Thorase Antibody, clone 9G7.1 is validated for use in Western Blotting for the detection of Thorase.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Thorase in 10 µg of human brain tissue lysate.

Quality

Evaluated by Western Blotting in human lymph node tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Thorase in 10 µg of human lymph node tissue lysate.

Target description

~41 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service