Skip to Content
Merck
All Photos(1)

Key Documents

MABF2092

Sigma-Aldrich

Anti-CD44 Antibody, clone Hermes-1

clone Hermes-1, from rat

Synonym(s):

CDw44, Epican, Extracellular matrix receptor III, ECMR-III, GP90, lymphocyte homing/adhesion receptor, HUTCH-I, Heparan sulfate proteoglycan, Hermes antigen, Hyaluronate receptor, Phagocytic glycoprotein 1, PGP-1, Phagocytic glycoprotein I, PGP-I

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€302.00

€116.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€302.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€116.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Hermes-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

General description

CD44 antigen (UniProt: P16070; also known as CDw44, Epican, Extracellular matrix receptor III, ECMR-III, GP90 lymphocyte homing/adhesion receptor, HUTCH-I, Heparan sulfate proteoglycan, Hermes antigen, Hyaluronate receptor, Phagocytic glycoprotein 1, PGP-1, Phagocytic glycoprotein I, PGP-I) is encoded by the CD44 (also known as LHR, MDU2, MDU3, MIC4) gene (Gene ID: 960) in human. CD44 is a single-pass type I membrane glycoprotein that is involved in cell-cell interactions, cell adhesion, and migration. It cell-cell and cell-matrix interactions are mediated through its affinity for hyaluronic acid (HA) and through other ligands. Adhesion with HA is considered to be important in cell migration, tumor growth, and progression. In cancer cells, it may play an important role in the formation of invadopodia. CD44 is synthesized with a signal peptide (aa 1-20), which is subsequently cleaved off to produce mature form. Its extracellular domain is localized to amino acids 21-649 and it has a short helical transmembrane domain (aa 650-670), and a cytoplasmic tail (aa 671-742). Nineteen different isoforms of CD44 are reported that are produced by alternative splicing.

Specificity

Clone hermes-1 is a rat monoclonal antibody that detects human CD44.

Immunogen

Human tonsil lymphocytes.

Application

Anti-CD44, clone Hermes-1, Cat. No. MABF2092, is a rat monoclonal antibody that detects human CD44 and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Immunoprecipitation.
Immunohistochemistry Analysis: A representative lot detected CD44 in Immunohistochemistry applications (Jalkanen, S.T., et. al. (1986). Eur J Immunol. 16(10):1195-202).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CD44 in Immunoprecipitation applications (Jalkanen, S.T., et. al. (1986). Eur J Immunol. 16(10):1195-202; Jalkanen, S., et. al. (1987). J Cell Biol. 105(2):983-90).

Flow Cytometry Analysis: A representative lot detected CD44 in Flow Cytometry applications (Jalkanen, S.T., et. al. (1986). Eur J Immunol. 16(10):1195-202).
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunohistochemistry in human skin and human sweat gland tissues.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected CD44 in human skin and human sweat gland tissues.

Target description

81.54 kDa calculated for isoform 1.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service