Skip to Content
Merck
All Photos(1)

Key Documents

HX0514

Sigma-Aldrich

Hyaluronidase

Synonym(s):

Hyaluronidase

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€233.00

€233.00


Estimated to ship on26 May 2025



Select a Size

Change View
1 PKG
€233.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.78

€233.00


Estimated to ship on26 May 2025


conjugate

conjugate (Glucosaminoglycan)

Quality Level

form

powder

specific activity

≥300 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

storage temp.

−20°C

General description

300 units/mg minimum, 50,000 units/vial. Suitable for biochemistry grade.

Specifications:Activity: 300NF U/mg min





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Hyaluronidase has been used:
  • to enzymatically remove extra-cellular matrix (ECM) to assess the lateral diffusion of the synaptic a-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptor (AMPAR)[1]
  • to study the effects of 4-methylumbelliferone (4MU)/ hyaluronidase (HLX)-mediated depletion of the CD44/hyaluronan complex on trastuzumab binding in JIMT1 breast cancer xenografts[2]
  • as a component of Hank′s balanced salt solution (HBSS) medium to digest fetal intestine tissue[3]

Biochem/physiol Actions

Hyaluronidase is an endoglycosidase that degrades hyaluronic acid (HA), a component of the extracellular matrix. It is used to enhance drug diffusion and reverse the effects of HA filler injections. Hyaluronidases dissolve the subcutaneous nodules and can correct excessive quantities of injected filler. It eliminates the cumulus-corona-oocyte complex formed during intracytoplasmic sperm injection. Hyaluronidase inhibits tissue damage after the extravasation of several substances. It is used for reducing edema treating vitreous hemorrhage.[4]

Unit Definition

One unit is based on the change in absorbency (turbidity) at 540 nm of an internal standard assayed concurrently with each lot. Internal standard replaces USP/NF reference no longer available.

Physical form

Dialyzed, lyophilized powder, not less than300 units per mg (dry weight).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maurizio Cavallini et al.
Aesthetic surgery journal, 33(8), 1167-1174 (2013-11-08)
Hyaluronidases, a family of enzymes that are able to degrade hyaluronic acid (HA), are employed in medicine to increase drug diffusion and reverse the effects of HA filler injections. Hyaluronidases are able to dissolve subcutaneous nodules or to correct excessive
Alexander D B Ross et al.
STAR protocols, 2(2), 100597-100597 (2021-06-26)
Here, we describe protocols for the preparation and dissociation of human fetal and pediatric intestinal tissue to a high-viability epithelial single-cell suspension. This epithelium-enriched single-cell suspension can then be used to generate single-cell RNA sequencing data as well as to
Sang Hak Lee et al.
eLife, 6 (2017-07-28)
Previous studies tracking AMPA receptor (AMPAR) diffusion at synapses observed a large mobile extrasynaptic AMPAR pool. Using super-resolution microscopy, we examined how fluorophore size and photostability affected AMPAR trafficking outside of, and within, post-synaptic densities (PSDs) from rats. Organic fluorescent
Patricia M R Pereira et al.
Molecular pharmaceutics, 17(1), 327-337 (2019-12-06)
Human epidermal growth factor receptor 2 (HER2) is a biomarker in breast cancer, and its overexpression is required to initiate therapies using HER2-targeted antibodies. Although trastuzumab is one of the most effective therapeutic antibodies in HER2-overexpressing breast cancer, a significant

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Questions

1–2 of 2 Questions  
  1. what is its source?

    1 answer
    1. Product HX0514 is from bovine testes.

      Helpful?

  2. What is the appropriate way to reconstitute this? In what medium and at what concentration? Is there an information sheet that provides this information?

    1 answer
    1. The enzyme can be reconstituted at 1mg/ml in 0.1M Sodium phosphate buffer, pH 5.3 with 0.15M sodium chloride. Stock solutions can be aliquoted and frozen for several months at -20C.

      Stock solutions prepared in water (30units/ml) are stable for up to 2 weeks at 4°C and stock solutions prepared in 500 mM acetate buffer, pH 6.0, are stable for up to 1 month at 4°C.

      It is recommended to make hyaluronidase solutions fresh. The lyophilized powder is very stable, at least 2 years when stored frozen -20C. This product does not contain a product information sheet.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service